Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR352906.1 | - | 1 - 2 | 138 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |