Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015280453.2 | XP_015135939.1 | 12 - 13 | 3357 | 119 | |
XM_025146897.1 | XP_025002665.1 | 14 - 15 | 1787 | 119 | ||
XM_417196.6 | XP_417196.1 | 13 - 14 | 1682 | 119 | ||
GenBank mRNA | BX929784.2 | - | 11 - 12 | 1081 | 119 | |
CR353417.1 | - | 2 - 3 | 911 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |