Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015280453.2 | XP_015135939.1 | 5 - 6 | 1085 | 103 | |
XM_015280465.2 | XP_015135951.1 | 5 - 6 | 968 | 103 | ||
XM_015280475.2 | XP_015135961.1 | 5 - 6 | 1085 | 103 | ||
XM_025146897.1 | XP_025002665.1 | 5 - 6 | 1085 | 103 | ||
XM_417196.6 | XP_417196.1 | 5 - 6 | 1012 | 103 | ||
GenBank mRNA | BX929784.2 | - | 3 - 4 | 411 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |