Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015280453.2 | XP_015135939.1 | 9 - 10 | 1499 | 62 | |
XM_015280465.2 | XP_015135951.1 | 8 - 9 | 1350 | 62 | ||
XM_015280475.2 | XP_015135961.1 | 8 - 9 | 1467 | 62 | ||
XM_025146897.1 | XP_025002665.1 | 9 - 10 | 1499 | 62 | ||
XM_417196.6 | XP_417196.1 | 8 - 9 | 1394 | 62 | ||
GenBank mRNA | BX929784.2 | - | 6 - 7 | 793 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |