Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277567.1 | NP_001264496.1 | 1 - 2 | 115 | 97 | |
GenBank mRNA | BU372764.1 | - | 1 - 2 | 85 | 97 | |
BU404580.1 | - | 1 - 2 | 115 | 97 | ||
BX931356.2 | - | 1 - 2 | 101 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |