Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277412.1 | NP_001264341.1 | 9 - 10 | 947 | 142 | |
GenBank mRNA | BU465115.1 | - | 4 - 5 | 289 | 142 | |
BX930716.1 | - | 9 - 10 | 899 | 142 | ||
BX932323.2 | - | 9 - 10 | 958 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |