Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146131.1 | NP_001139603.1 | 1 - 2 | 398 | 58 | |
GenBank mRNA | BX932928.2 | - | 1 - 2 | 418 | 58 | |
FN178490.1 | - | 1 - 2 | 398 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |