Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277375.1 | NP_001264304.1 | 4 - 5 | 505 | 85 | |
XM_015288649.2 | XP_015144135.1 | 4 - 5 | 511 | 85 | ||
GenBank mRNA | BU109911.1 | - | 2 - 3 | 205 | 85 | |
BX931374.1 | - | 4 - 5 | 494 | 85 | ||
BX934375.2 | - | 4 - 5 | 446 | 85 | ||
BX935049.1 | - | 4 - 5 | 486 | 85 | ||
CD739203.1 | - | 4 - 5 | 505 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |