Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197245.1 | NP_001184174.1 | 2 - 3 | 282 | 67 | |
GenBank mRNA | BX935038.2 | - | 2 - 3 | 282 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |