Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001305.3 | NP_001001305.1 | 3 | 2134 | 118 | |
XM_015295121.2 | XP_015150607.1 | 3 | 3316 | 118 | ||
XM_015295122.2 | XP_015150608.1 | 4 | 3046 | 118 | ||
XM_025141547.1 | XP_024997315.1 | 4 | 2264 | 118 | ||
GenBank mRNA | BX933423.1 | - | 1 | 640 | 116 | |
BX935425.2 | - | 1 | 554 | 117 |
Gene Symbol: METTL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642337.4 | XP_003642385.1 | 5 | 1492 | 118 | |
XM_004946111.3 | XP_004946168.1 | 4 | 1363 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |