Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277059.2 | XP_015132545.1 | 3 - 4 | 322 | 59 | |
GenBank mRNA | BX935736.2 | - | 3 - 4 | 311 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |