Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277590.1 | NP_001264519.1 | 1 - 2 | 45 | 71 | |
GenBank mRNA | BM440029.1 | - | 1 - 2 | 40 | 71 | |
BX540414.3 | - | 1 - 2 | 52 | 71 | ||
BX950422.2 | - | 1 - 2 | 38 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |