Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277590.1 | NP_001264519.1 | 6 - 7 | 627 | 96 | |
GenBank mRNA | BU380556.1 | - | 5 - 6 | 612 | 96 | |
BX950422.2 | - | 6 - 7 | 619 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |