Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282541.2 | XP_015138027.2 | 53 - 54 | 8607 | 133 | |
XM_015282543.2 | XP_015138029.2 | 52 - 53 | 8439 | 133 | ||
XM_025147818.1 | XP_025003586.1 | 53 - 54 | 8610 | 133 | ||
XM_025147819.1 | XP_025003587.1 | 52 - 53 | 8498 | 133 | ||
XM_025147820.1 | XP_025003588.1 | 53 - 54 | 8607 | 133 | ||
XM_025147821.1 | XP_025003589.1 | 49 - 50 | 7973 | 133 | ||
XM_025147822.1 | XP_025003590.1 | 52 - 53 | 8571 | 133 | ||
XM_025147824.1 | XP_025003592.1 | 52 - 53 | 8442 | 133 | ||
GenBank mRNA | CR385783.1 | - | 3 - 4 | 473 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |