Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278005.1 | NP_001264934.1 | 1 - 2 | 189 | 56 | |
GenBank mRNA | CR385804.1 | - | 1 - 2 | 177 | 56 | |
CV041818.1 | - | 1 - 2 | 189 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |