Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945285.3 | XP_004945342.2 | 5 - 6 | 503 | 99 | |
XM_414903.6 | XP_414903.3 | 5 - 6 | 488 | 99 | ||
GenBank mRNA | CR386453.1 | - | 5 - 6 | 361 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |