Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299083.2 | XP_015154569.1 | 560 | 98 | ||
XM_015299085.2 | XP_015154571.1 | 560 | 98 | |||
XM_417963.6 | XP_417963.3 | 560 | 98 | |||
GenBank mRNA | BX929280.1 | - | 199 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus |