Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277440.1 | NP_001264369.1 | 1 - 2 | 135 | 102 | |
GenBank mRNA | BX929424.1 | - | 1 - 2 | 97 | 102 | |
CN237805.1 | - | 1 - 2 | 135 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |