Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277054.2 | XP_015132540.1 | 2 - 3 | 144 | 95 | |
XM_025153690.1 | XP_025009458.1 | 2 - 3 | 417 | 95 | ||
GenBank mRNA | BX930373.1 | - | 2 - 3 | 320 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |