Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149901.1 | XP_025005669.1 | 7 - 8 | 892 | 149 | |
XM_025149902.1 | XP_025005670.1 | 7 - 8 | 892 | 149 | ||
XM_025149903.1 | XP_025005671.1 | 7 - 8 | 892 | 149 | ||
XM_025149904.1 | XP_025005672.1 | 7 - 8 | 892 | 149 | ||
XM_025149906.1 | XP_025005674.1 | 7 - 8 | 892 | 149 | ||
XM_025149907.1 | XP_025005675.1 | 7 - 8 | 892 | 149 | ||
XM_025149908.1 | XP_025005676.1 | 7 - 8 | 892 | 149 | ||
XM_025149909.1 | XP_025005677.1 | 7 - 8 | 892 | 149 | ||
XM_420764.6 | XP_420764.2 | 7 - 8 | 892 | 149 | ||
GenBank mRNA | BX930771.1 | - | 7 - 8 | 690 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |