Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277375.1 | NP_001264304.1 | 1 - 2 | 89 | 87 | |
XM_015288649.2 | XP_015144135.1 | 1 - 2 | 95 | 87 | ||
GenBank mRNA | BX931374.1 | - | 1 - 2 | 78 | 87 | |
CD739203.1 | - | 1 - 2 | 89 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |