Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415401.6 | XP_415401.2 | 5 - 6 | 544 | 63 | |
GenBank mRNA | BX934525.1 | - | 5 - 6 | 539 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |