Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025146941.1 | XP_025002709.1 | 14 - 15 | 2444 | 100 | |
GenBank mRNA | BX934563.1 | - | 1 - 2 | 151 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |