Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015292675.2 | XP_015148161.2 | 12 - 13 | 2680 | 76 | |
GenBank mRNA | CR386582.1 | - | 1 - 2 | 213 | 76 | |
CR391412.1 | - | 1 - 2 | 199 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |