Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015292675.2 | XP_015148161.2 | 13 - 14 | 2743 | 84 | |
GenBank mRNA | CR386582.1 | - | 2 - 3 | 276 | 84 | |
CR391412.1 | - | 2 - 3 | 262 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |