Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422010.6 | XP_422010.1 | 4 - 5 | 2264 | 133 | |
GenBank mRNA | CR405927.1 | - | 4 - 5 | 641 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |