Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR406158.1 | - | 4 - 5 | 820 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |