Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419897.6 | XP_419897.5 | 3 | 680 | 132 | |
GenBank mRNA | BX933957.2 | - | 3 | 649 | 132 | |
CR406498.1 | - | 3 | 649 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |