Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006313.2 | NP_001006313.2 | 1 - 2 | 217 | 126 | |
XM_015297665.2 | XP_015153151.1 | 2 - 3 | 256 | 126 | ||
XM_015297666.2 | XP_015153152.1 | 2 - 3 | 810 | 126 | ||
XM_025142994.1 | XP_024998762.1 | 2 - 3 | 235 | 126 | ||
XM_025142995.1 | XP_024998763.1 | 2 - 3 | 254 | 126 | ||
GenBank mRNA | AJ720022.1 | - | 2 - 3 | 254 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |