Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006329.1 | NP_001006329.1 | 3 - 4 | 561 | 90 | |
GenBank mRNA | AJ721086.1 | - | 3 - 4 | 561 | 90 | |
BX931317.2 | - | 4 - 5 | 344 | 90 | ||
CR354048.1 | - | 1 - 2 | 286 | 90 | ||
CR386397.1 | - | 2 - 3 | 431 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |