Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006334.1 | NP_001006334.1 | 7 - 8 | 821 | 75 | |
XM_015299602.2 | XP_015155088.1 | 8 - 9 | 1065 | 75 | ||
XM_015299603.2 | XP_015155089.1 | 8 - 9 | 772 | 75 | ||
XM_015299604.2 | XP_015155090.1 | 8 - 9 | 815 | 75 | ||
XM_025143978.1 | XP_024999746.1 | 6 - 7 | 632 | 75 | ||
GenBank mRNA | AJ720128.1 | - | 7 - 8 | 821 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |