Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006335.2 | NP_001006335.2 | 10 - 11 | 1128 | 92 | |
XM_015299606.2 | XP_015155092.1 | 10 - 11 | 1135 | 92 | ||
GenBank mRNA | AJ719694.1 | - | 10 - 11 | 1128 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |