Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006349.1 | NP_001006349.1 | 10 - 11 | 1075 | 75 | |
GenBank mRNA | AJ719829.1 | - | 10 - 11 | 1075 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |