Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006382.1 | NP_001006382.1 | 8 - 9 | 1279 | 103 | |
GenBank mRNA | AJ719803.1 | - | 8 - 9 | 1279 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |