Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006446.1 | NP_001006446.1 | 2 - 3 | 533 | 79 | |
XM_015276238.2 | XP_015131724.1 | 2 - 3 | 536 | 79 | ||
GenBank mRNA | AJ720952.1 | - | 2 - 3 | 533 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |