Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006454.1 | NP_001006454.1 | 2 - 3 | 248 | 110 | |
XM_015285436.2 | XP_015140922.1 | 2 - 3 | 365 | 110 | ||
XM_025149692.1 | XP_025005460.1 | 2 - 3 | 584 | 110 | ||
GenBank mRNA | AJ720325.1 | - | 2 - 3 | 248 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |