Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006477.1 | NP_001006477.1 | 4 - 5 | 577 | 77 | |
XM_025150723.1 | XP_025006491.1 | 4 - 5 | 571 | 77 | ||
GenBank mRNA | AJ719612.1 | - | 4 - 5 | 577 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |