Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006481.1 | NP_001006481.1 | 3 - 4 | 452 | 89 | |
XM_015287388.2 | XP_015142874.1 | 3 - 4 | 485 | 89 | ||
GenBank mRNA | AJ719909.1 | - | 3 - 4 | 452 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |