Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204558.1 | NP_989889.1 | 3 - 4 | 454 | 138 | |
XM_025142159.1 | XP_024997927.1 | 3 - 4 | 755 | 138 | ||
GenBank mRNA | AF380350.1 | - | 3 - 4 | 454 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |