Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204558.1 | NP_989889.1 | 6 - 7 | 1026 | 76 | |
XM_025142159.1 | XP_024997927.1 | 6 - 7 | 1327 | 76 | ||
GenBank mRNA | AF380350.1 | - | 6 - 7 | 1026 | 76 | |
CR385416.1 | - | 1 - 2 | 135 | 76 | ||
CR387664.1 | - | 3 - 4 | 166 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |