Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204585.1 | NP_989916.1 | 3 - 4 | 334 | 73 | |
XM_015296819.2 | XP_015152305.1 | 3 - 4 | 273 | 73 | ||
GenBank mRNA | AB076264.1 | - | 3 - 4 | 334 | 73 | |
HM012714.1 | - | 2 - 3 | 213 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |