Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204602.1 | NP_989933.1 | 2 - 3 | 295 | 112 | |
XM_015293587.2 | XP_015149073.1 | 3 - 4 | 426 | 112 | ||
GenBank mRNA | AF077369.1 | - | 2 - 3 | 303 | 112 | |
AF420468.1 | - | 2 - 3 | 295 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |