Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204603.1 | NP_989934.1 | 5 - 6 | 2012 | 76 | |
XM_015289377.2 | XP_015144863.1 | 6 - 7 | 2125 | 76 | ||
XM_025152088.1 | XP_025007856.1 | 3 - 4 | 1830 | 76 | ||
GenBank mRNA | AJ317961.1 | - | 5 - 6 | 2012 | 76 | |
AJ719471.1 | - | 3 - 4 | 1660 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |