Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204646.1 | NP_989977.1 | 2 - 3 | 1658 | 86 | |
XM_015291636.2 | XP_015147122.1 | 2 - 3 | 1593 | 86 | ||
XM_015291637.1 | XP_015147123.1 | 2 - 3 | 1545 | 86 | ||
XM_015291639.2 | XP_015147125.1 | 2 - 3 | 1593 | 86 | ||
XM_015291640.2 | XP_015147126.1 | 2 - 3 | 1593 | 86 | ||
XM_025153307.1 | XP_025009075.1 | 2 - 3 | 1576 | 86 | ||
GenBank mRNA | AJ306928.1 | - | 2 - 3 | 1658 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |