Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204671.1 | NP_990002.1 | 5 - 6 | 501 | 100 | |
XM_015293731.2 | XP_015149217.1 | 5 - 6 | 539 | 100 | ||
GenBank mRNA | AF253514.1 | - | 5 - 6 | 501 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |