Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204684.2 | NP_990015.2 | 1 | 536 | 64 | |
GenBank mRNA | AJ292363.1 | - | 1 | 549 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |