Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204687.2 | NP_990018.1 | 3 - 4 | 544 | 57 | |
XM_015297949.2 | XP_015153435.1 | 4 - 5 | 733 | 57 | ||
XM_015297950.2 | XP_015153436.1 | 5 - 6 | 800 | 57 | ||
XM_015297951.2 | XP_015153437.1 | 4 - 5 | 1294 | 57 | ||
XM_025143171.1 | XP_024998939.1 | 5 - 6 | 807 | 57 | ||
XM_025143172.1 | XP_024998940.1 | 4 - 5 | 1162 | 57 | ||
GenBank mRNA | AF292934.1 | - | 3 - 4 | 544 | 57 | |
Y08170.2 | - | 521 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |