Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204696.1 | NP_990027.1 | 2 - 3 | 684 | 162 | |
XM_015277235.2 | XP_015132721.1 | 3 - 4 | 1977 | 162 | ||
GenBank mRNA | D86333.1 | - | 2 - 3 | 684 | 162 | |
KY747398.1 | - | 2 - 3 | 480 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |