Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204812.2 | NP_990143.2 | 3 - 4 | 254 | 79 | |
XM_015280411.2 | XP_015135897.1 | 3 - 4 | 695 | 79 | ||
XM_025144652.1 | XP_025000420.1 | 2 - 3 | 2127 | 79 | ||
XM_025144653.1 | XP_025000421.1 | 2 - 3 | 2123 | 79 | ||
GenBank mRNA | X83739.2 | - | 254 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |