Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204818.2 | NP_990149.1 | 2 - 3 | 146 | 97 | |
XM_015284816.2 | XP_015140302.1 | 3 - 4 | 245 | 97 | ||
GenBank mRNA | AF133251.1 | - | 2 - 3 | 149 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |